missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-7-azaindole, 97%

Catalog No. AC437430050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437430050 5 g Glass bottle
AC437430010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437430050 Supplier Thermo Scientific Chemicals Supplier No. 437430050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 183208-35-7 | C7H5BrN2 | 197.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 183208-35-7
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD06659677
InChI Key LPTVWZSQAIDCEB-UHFFFAOYSA-N
PubChem CID 10307932
IUPAC Name 5-bromo-1H-pyrrolo[2,3-b]pyridine
SMILES BrC1=CN=C2NC=CC2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 179.7°C to 180.7°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Synonym 5-bromo-7-azaindole, 5-bromo-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 5-bromo, 5-bromo azaindole, 3-bromo-7h-pyrrolo 2,3-b pyridine, 5-bromo-1h-pyrrolo 2 , 3-b pyridine, 5-bromopyrrolo 2,3-b pyridine, 5-bromo,7-azaindole
Formula Weight 197.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-7-azaindole
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

RUO – Research Use Only