missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-7-azaindole, 96%, Thermo Scientific Chemicals

Catalog No. AAH5004003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5004003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5004003 Supplier Thermo Scientific Chemicals Supplier No. H5004003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 183208-35-7
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD06659677
InChI Key LPTVWZSQAIDCEB-UHFFFAOYSA-N
PubChem CID 10307932
SMILES BrC1=CN=C2NC=CC2=C1

Specifications

Melting Point 178°C to 179°C
Quantity 1 g
Synonym 5-bromo-7-azaindole, 5-bromo-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 5-bromo, 5-bromo azaindole, 3-bromo-7h-pyrrolo 2,3-b pyridine, 5-bromo-1h-pyrrolo 2 , 3-b pyridine, 5-bromopyrrolo 2,3-b pyridine, 5-bromo,7-azaindole
IUPAC Name 5-bromo-1H-pyrrolo[2,3-b]pyridine
Formula Weight 197.04
Percent Purity 96%
Chemical Name or Material 5-Bromo-7-azaindole
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only