missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-6-hydroxynicotinic Acid 98.0+%, TCI America™

Catalog No. B51931G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51931G 1 g
B51935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51931G Supplier TCI America Supplier No. B51931G

Chemical Identifiers

CAS 41668-13-7
Molecular Formula C6H4BrNO3
Molecular Weight (g/mol) 218.006
MDL Number MFCD08235173
InChI Key PQDLYKZCJBGXPQ-UHFFFAOYSA-N
Synonym 5-Bromo-6-hydroxy-3-pyridinecarboxylic Acid
PubChem CID 2764314
IUPAC Name 5-bromo-6-oxo-1H-pyridine-3-carboxylic acid
SMILES C1=C(C(=O)NC=C1C(=O)O)Br

Specifications

Melting Point 296°C
Color White-Yellow
Quantity 1 g
Formula Weight 218.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-6-hydroxynicotinic Acid