missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-β-D-glucuronide Sodium Salt, MP Biomedicals™

Catalog No. ICN19392505
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19392505 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19392505 Supplier MP Biomedicals Inc Supplier No. 0219392505

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A β-glucuronidase substrate which forms an intense blue precipitate upon enzymatic action. Used for the detection of the GUS gene in bacterial colonies and in histochemical applications

Chemical Identifiers

CAS 129541-41-9
Molecular Formula C14H12BrClNNaO7
Molecular Weight (g/mol) 444.594
InChI Key IBLSVGDGSKUDCT-ILIJQVQCSA-M
Synonym x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt
PubChem CID 53384407
IUPAC Name sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+]

Specifications

Color White
Assay Percent Range ≥98%
Quantity 5 mg
Formula Weight 444.6
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-β-D-glucuronide Sodium Salt