missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-4-chloro-3-indolyl-βD-galactopyranoside, Promega
Description
- Quality tested for use with the pGEM(R)-Z Vectors in a chromogenicity assay
- Supplied at a concentration of 50mg/mL in dimethylformamide, 2.0ml/vial
Specifications
Specifications
| Quantity | 100 mg |
| Formula Weight | 408.63 |
| Grade | Analytical |
| Chemical Name or Material | 5-Bromo-4-Chloro-3-Indolyl-Beta-D-Galactopyranoside |
Safety and Handling
| Recommended Storage | −30 to −10°C |