missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215000105

 View more versions of this product

Catalog No. ICN15000105


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside
7240-90-6
0.994
MFCD00005666
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
408.629
CHEBI:75055
Crystalline Powder
For HPLC analysis
White
C14H15BrClNO6
5 mg
OPIFSICVWOWJMJ-AEOCFKNESA-N
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
65181
408.64

Chemical Identifiers

7240-90-6
408.629
OPIFSICVWOWJMJ-AEOCFKNESA-N
65181
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C14H15BrClNO6
MFCD00005666
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
CHEBI:75055
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 230-640-8

Recommended Storage : Store at -20°C, desiccate, protect from light .

SDS
Documents

Documents

Promotions

Promotions