missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside, ≥98%, Molecular Biology Reagent, MP Biomedicals™

Catalog No. ICN19481110
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19481110 10 mg
ICN19481180 100 mg
ICN19481190 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19481110 Supplier MP Biomedicals Inc Supplier No. 0219481110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside is used as indigogenic substrate for β-galactosidase, for detection of β-galactosidase-positive clones, and the identification of lac and bacterial colonies or phage plaques

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
MDL Number MFCD00005666
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

Color White
Assay Percent Range ≥98%
Quantity 10 mg
Formula Weight 408.6
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material 5-Bromo-4-Chloro-3-Indoyl-β-D-Galactopyranoside, FTIR