missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%

Substrate for beta→-D-glucosidase | CAS: 15548-60-4 | C14H15BrClNO6 | 408.63 g/mol

$117.56 - $215.37

Chemical Identifiers

CAS 15548-60-4
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.63
MDL Number MFCD00063690
InChI Key OPIFSICVWOWJMJ-LNNRFACYSA-N
Synonym x-glc, x-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucopyranoside, 5-bromo-4-chloroindol-3-yl-beta-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl b-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-glucoside, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, indoxyl-glucoside
PubChem CID 84982
ChEBI CHEBI:75424
IUPAC Name (2S,3R,4S,5S,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)[C@H](O)[C@@H](O)[C@@H]1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65677ME
View Documents
Thermo Scientific Chemicals
J65677ME
500 mg
Each for $117.56
Only null left
 
AAJ6567703
View Documents
Thermo Scientific Chemicals
J6567703
1 g
Each for $215.37
Only null left
 
Description

Description

Substrate for beta--D-glucosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15548-60-4
408.63
OPIFSICVWOWJMJ-LNNRFACYSA-N
84982
(2S,3R,4S,5S,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C14H15BrClNO6
MFCD00063690
x-glc, x-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucopyranoside, 5-bromo-4-chloroindol-3-yl-beta-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl b-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-glucoside, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, indoxyl-glucoside
CHEBI:75424
OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

15548-60-4
C14H15BrClNO6
500 mg
x-glc, x-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucopyranoside, 5-bromo-4-chloroindol-3-yl-beta-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl b-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-glucoside, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, indoxyl-glucoside
OPIFSICVWOWJMJ-LNNRFACYSA-N
(2S,3R,4S,5S,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
84982
408.63
Crystalline Powder
White
MFCD00063690
1552603
Soluble in DMF or DMSO
OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)[C@H](O)[C@@H](O)[C@@H]1O
408.63
CHEBI:75424
99%
5-Bromo-4-chloro-3-indolyl beta-D-glucoside
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 239-603-0

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only