missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%

Catalog No. AAJ65677ME
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65677ME 500 mg
AAJ6567703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65677ME Supplier Thermo Scientific Chemicals Supplier No. J65677ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Substrate for beta→-D-glucosidase | CAS: 15548-60-4 | C14H15BrClNO6 | 408.63 g/mol

Substrate for beta--D-glucosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15548-60-4
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.63
MDL Number MFCD00063690
InChI Key OPIFSICVWOWJMJ-LNNRFACYSA-N
Synonym x-glc, x-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucopyranoside, 5-bromo-4-chloroindol-3-yl-beta-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl b-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-glucoside, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, indoxyl-glucoside
PubChem CID 84982
ChEBI CHEBI:75424
IUPAC Name (2S,3R,4S,5S,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Color White
Quantity 500 mg
Beilstein 1552603
Solubility Information Soluble in DMF or DMSO
Formula Weight 408.63
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl beta-D-glucoside
EINECSNumber 239-603-0
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only