missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-4-chloro-3-indolyl beta-D-Glucopyranoside 98.0+%, TCI America™
$69.31 - $192.52
Chemical Identifiers
| CAS | 15548-60-4 |
|---|---|
| Molecular Formula | C14H15BrClNO6 |
| Molecular Weight (g/mol) | 408.63 |
| MDL Number | MFCD00063690 |
| InChI Key | OPIFSICVWOWJMJ-LNNRFACYSA-N |
| Synonym | x-glc, x-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucopyranoside, 5-bromo-4-chloroindol-3-yl-beta-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl b-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-glucoside, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, indoxyl-glucoside |
| PubChem CID | 84982 |
| ChEBI | CHEBI:75424 |
| IUPAC Name | (2S,3R,4S,5S,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
| SMILES | OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)[C@H](O)[C@@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5393200MG
|
TCI America
B5393200MG |
200 mg |
Each for $69.31
|
|
|||||
|
B53931G
|
TCI America
B53931G |
1 g |
Each for $192.52
|
|
|||||
Chemical Identifiers
| 15548-60-4 | |
| 408.63 | |
| OPIFSICVWOWJMJ-LNNRFACYSA-N | |
| 84982 | |
| (2S,3R,4S,5S,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
| C14H15BrClNO6 | |
| MFCD00063690 | |
| x-glc, x-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucopyranoside, 5-bromo-4-chloroindol-3-yl-beta-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl b-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-glucoside, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, indoxyl-glucoside | |
| CHEBI:75424 | |
| OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)[C@H](O)[C@@H](O)[C@@H]1O |
Specifications
| 15548-60-4 | |
| White-Yellow | |
| MFCD00063690 | |
| x-glc, x-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucopyranoside, 5-bromo-4-chloroindol-3-yl-beta-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl b-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-glucoside, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, indoxyl-glucoside | |
| OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)[C@H](O)[C@@H](O)[C@@H]1O | |
| 408.63 | |
| CHEBI:75424 | |
| ≥98.0% (HPLC) | |
| 5-Bromo-4-chloro-3-indolyl beta-D-Glucopyranoside [for Biochemical Research] |
| 243°C | |
| C14H15BrClNO6 | |
| 200 mg | |
| OPIFSICVWOWJMJ-LNNRFACYSA-N | |
| (2S,3R,4S,5S,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 84982 | |
| 408.63 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)