missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-4-chloro-3-indolyl beta-D-Glucopyranoside 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 243°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 408.63 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Bromo-4-chloro-3-indolyl beta-D-Glucopyranoside [for Biochemical Research] |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer (-20 °C) |