missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99+%, for molecular biology, DNAse, RNAse and Protease free
CAS: 7240-90-6 | C14H15BrClNO6 | 408.63 g/mol
$193.06
Chemical Identifiers
| CAS | 7240-90-6 |
|---|---|
| Molecular Formula | C14H15BrClNO6 |
| Molecular Weight (g/mol) | 408.63 |
| MDL Number | MFCD00005666 |
| InChI Key | OPIFSICVWOWJMJ-AEOCFKNESA-N |
| Synonym | x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal |
| PubChem CID | 65181 |
| ChEBI | CHEBI:75055 |
| IUPAC Name | (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
| SMILES | C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC327241000
|
Thermo Scientific Chemicals
327241000 |
100 mg | Glass bottle |
Each for $193.06
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7240-90-6 | |
| 408.63 | |
| OPIFSICVWOWJMJ-AEOCFKNESA-N | |
| 65181 | |
| (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
| C14H15BrClNO6 | |
| MFCD00005666 | |
| x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal | |
| CHEBI:75055 | |
| C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br |
Specifications
| 7240-90-6 | |
| 100.0 | |
| White | |
| 99+% | |
| MFCD00005666 | |
| − 64.00 | |
| OPIFSICVWOWJMJ-AEOCFKNESA-N | |
| (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 65181 | |
| 408.63 | |
| Molecular Biology | |
| 5-Bromo-4-chloro-3-indolyl-β-D-galactoside, For molecular biology |
| 99.0 | |
| 230.0°C | |
| Authentic | |
| C14H15BrClNO6 | |
| 15, 10272 | |
| x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal | |
| C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br | |
| 408.63 | |
| CHEBI:75055 | |
| 99+% | |
| Powder |
Safety and Handling
EINECSNumber : 230-640-8
RUO – Research Use Only