missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99+%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. AC327245000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327245000 500 mg Glass bottle
AC327241000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327245000 Supplier Thermo Scientific Chemicals Supplier No. 327245000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7240-90-6 | C14H15BrClNO6 | 408.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.63
MDL Number MFCD00005666
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 230.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 500 mg
Merck Index 15, 10272
Specific Rotation Condition - 64.00 (20.00°C c=1,H2O/DMF (1/1))
Specific Rotation − 64.00
Formula Weight 408.63
Percent Purity 99+%
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-β-D-galactoside, For molecular biology
Show More Show Less
EINECSNumber 230-640-8

RUO – Research Use Only