missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-4-chloro-3-indolyl beta-D-Galactopyranoside 98.0+%, TCI America™
$94.93 - $282.90
Chemical Identifiers
| CAS | 7240-90-6 |
|---|---|
| Molecular Formula | C14H15BrClNO6 |
| Molecular Weight (g/mol) | 408.629 |
| MDL Number | MFCD00005666 |
| InChI Key | OPIFSICVWOWJMJ-AEOCFKNESA-N |
| Synonym | x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal |
| PubChem CID | 65181 |
| ChEBI | CHEBI:75055 |
| IUPAC Name | (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
| SMILES | C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
B3201200MG
|
N/A | 200 mg |
Each for $94.93
|
|
|||||
|
B32011G
|
N/A | 1 g |
Each for $282.90
|
|
|||||
Chemical Identifiers
| 7240-90-6 | |
| 408.629 | |
| OPIFSICVWOWJMJ-AEOCFKNESA-N | |
| 65181 | |
| (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
| C14H15BrClNO6 | |
| MFCD00005666 | |
| x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal | |
| CHEBI:75055 | |
| C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br |
Specifications
| 7240-90-6 | |
| C14H15BrClNO6 | |
| x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal | |
| C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br | |
| 408.629 | |
| CHEBI:75055 | |
| ≥98.0% (HPLC,N) | |
| 5-Bromo-4-chloro-3-indolyl beta-D-Galactopyranoside [for Biochemical Research] |
| White | |
| MFCD00005666 | |
| OPIFSICVWOWJMJ-AEOCFKNESA-N | |
| (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 65181 | |
| 408.63 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer