missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside, 98+%

Catalog No. p-7025521
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21034MF 50 mg
AAB21034MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB21034MF Supplier Thermo Scientific Chemicals Supplier No. B21034MF
Only null left

CAS: 7240-90-6 | C14H15BrClNO6 | 408.629 g/mol

It is a substrate for beta-galactosidase, which cleaves the glycosidic bond to give 5-bromo-4-chloro-3-hydroxy-1H-indole, which immediately dimerises to give an intensely blue product. It is also used to detect activity of this enzyme in histochemistry and bacteriology. It is employed as a colorless, endogenous or exogenous pigment precursors that may be transformed by biological mechanisms into colored compounds. They are used in biochemical assays and in diagnosis as indicators, particularly in the form of enzyme substrates. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a substrate for beta-galactosidase, which cleaves the glycosidic bond to give 5-bromo-4-chloro-3-hydroxy-1H-indole, which immediately dimerises to give an intensely blue product. It is also used to detect activity of this enzyme in histochemistry and bacteriology. It is employed as a colorless, endogenous or exogenous pigment precursors that may be transformed by biological mechanisms into colored compounds. They are used in biochemical assays and in diagnosis as indicators, particularly in the form of enzyme substrates. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

Solubility
Soluble in 100 mM in DMSO.

Notes
Moisture & Light sensitive. Store away from oxidizing agents, heat, water/moisture and light. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
MDL Number MFCD00005666
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

Melting Point ∼236°C (decomposition)
Quantity 50 mg
Beilstein 1402009
Sensitivity Light and moisture sensitive
Solubility Information Soluble in 100 mM in DMSO.
Optical Rotation −62° (c=1 in 50% DMF)
Formula Weight 408.64
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 230-640-8
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only