missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside

Substrate for beta-galactosidase | CAS: 107021-38-5 | C14H15BrClNO6 | 408.63 g/mol

Supplier:  Thermo Scientific Chemicals J6015103

Quantity 1 g

 View more versions of this product

Catalog No. AAJ6015103


Only null left

Description

Description

Substrate for beta-galactosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

107021-38-5
408.63
OPIFSICVWOWJMJ-UHFFFAOYNA-N
10173103
OCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O
C14H15BrClNO6
MFCD00063780
x-alpha-gal, 5-bromo-4-chloro-3-indolyl-a-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl a-d-galactopyranoside, 5-bromo-4-chloroindoxyl alpha-d-galactoside, 5-bromo-4-chloro-3-indolyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactopyranoside, 2r,3r,4s,5r,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, x-alpha-d-galactoside, 5-bromo-4-chloro-3-indoxyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactoside
CHEBI:75506

Specifications

5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
White
1 g
x-alpha-gal, 5-bromo-4-chloro-3-indolyl-a-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl a-d-galactopyranoside, 5-bromo-4-chloroindoxyl alpha-d-galactoside, 5-bromo-4-chloro-3-indolyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactopyranoside, 2r,3r,4s,5r,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, x-alpha-d-galactoside, 5-bromo-4-chloro-3-indoxyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactoside
OCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O
408.63
CHEBI:75506
Powder
107021-38-5
C14H15BrClNO6
MFCD00063780
OPIFSICVWOWJMJ-UHFFFAOYNA-N
(2R,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
10173103
408.63
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions