missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-4-chloro-3-idolyl-α-D-galactopyranoside, MP Biomedicals™
Specifications
Specifications
| Color | White |
| Assay Percent Range | >99% |
| Quantity | 25 mg |
| Formula Weight | 408.63 |
| Grade | Research, NA |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Bromo-4-chloro-3-idolyl-α-D-galactopyranoside |