missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-pyridinemethanol 98.0+%, TCI America™

Catalog No. p-7122982
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34931G 1 g
B34935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34931G Supplier TCI America Supplier No. B34931G

Chemical Identifiers

CAS 37669-64-0
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD03265757
InChI Key WDVDHJLKXYCOFS-UHFFFAOYSA-N
Synonym 5-bromopyridin-3-yl methanol, 5-bromo-3-pyridinemethanol, 3-bromo-5-hydroxymethylpyridine, 5-bromo-3-hydroxymethylpyridine, 5-bromo-3-pyridinyl methanol, 5-bromo-pyridin-3-yl-methanol, 3-pyridinemethanol, 5-bromo, 5-bromo-3-pyridyl methan-1-ol, 3-bromo-5-hydroxymethyl pyridine, 3-bromo-pyridin-5-yl methanol
PubChem CID 2784733
IUPAC Name (5-bromopyridin-3-yl)methanol
SMILES OCC1=CC(Br)=CN=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-3-pyridinemethanol