missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. p-7122979
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34901G 1 g
B34905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34901G Supplier TCI America Supplier No. B34901G

Chemical Identifiers

CAS 113118-81-3
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
MDL Number MFCD03265758
InChI Key NGUVGKAEOFPLDT-UHFFFAOYSA-N
Synonym 5-bromonicotinaldehyde, 5-bromo-3-pyridinecarboxaldehyde, 5-bromo-3-formylpyridine, 3-bromopyridine-5-carboxaldehyde, 5-bromopyridine-3-carboxaldehyde, 5-bromo-pyridine-3-carbaldehyde, 3-bromo-5-formylpyridine, 3-pyridinecarboxaldehyde, 5-bromo, 3-bromopyridine-5-carbaldehyde, 5-bromo-3-pyridinecarbaldehyde
PubChem CID 2784734
IUPAC Name 5-bromopyridine-3-carbaldehyde
SMILES C1=C(C=NC=C1Br)C=O

Specifications

Melting Point 98°C
Quantity 1 g
Formula Weight 186.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-3-pyridinecarboxaldehyde