missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-pyridinecarbonitrile 98.0+%, TCI America™

Catalog No. B37131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37131G 1 g
B37135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37131G Supplier TCI America Supplier No. B37131G
Only null left

Chemical Identifiers

CAS 35590-37-5
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.01
MDL Number MFCD00174363
InChI Key FTFFHWWIPOQCBC-UHFFFAOYSA-N
Synonym 5-bromonicotinonitrile, 3-bromo-5-cyanopyridine, 5-bromo-3-cyanopyridine, 3-cyano-5-bromopyridine, 3-pyridinecarbonitrile, 5-bromo, 3-bromo-5-cyano-pyridine, 5-bromo-3-pyridinecarbonitrile, pubchem7702, acmc-209ihz, ksc492g7p
PubChem CID 736793
IUPAC Name 5-bromopyridine-3-carbonitrile
SMILES BrC1=CN=CC(=C1)C#N

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 183.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-3-pyridinecarbonitrile