missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-3-pyridineacetic acid, 98+%
Description
5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 182°C to 185°C |
| Quantity | 1 g |
| Beilstein | 388264 |
| Solubility Information | Soluble in water. |
| Formula Weight | 216.03 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 5-Bromo-3-pyridineacetic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only