missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-pyridineacetic acid, 98+%

Catalog No. AAL1357903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1357903 1 g
AAL1357906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1357903 Supplier Thermo Scientific Chemicals Supplier No. L1357903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39891-12-8 | C7H6BrNO2 | 216.034 g/mol

5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 39891-12-8
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00829308
InChI Key UETIDNDXXGCJCE-UHFFFAOYSA-N
Synonym 5-bromo-3-pyridylacetic acid, 2-5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridineacetic acid, 3-pyridineacetic acid, 5-bromo, 2-5-bromo-3-pyridyl acetic acid, 5-bromo-3-pyridylaceticacid, 5-bromo-pyridin-3-yl-acetic acid, 5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridinyl acetic acid, 2-5-bromopyridin-3-yl acetic acid
PubChem CID 2802539
IUPAC Name 2-(5-bromopyridin-3-yl)acetic acid
SMILES C1=C(C=NC=C1Br)CC(=O)O

Specifications

Melting Point 182°C to 185°C
Quantity 1 g
Beilstein 388264
Solubility Information Soluble in water.
Formula Weight 216.03
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-3-pyridineacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only