missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-methoxyquinoline, 96%

Catalog No. p-7036601
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54919MD 250 mg
AAH5491903 1 g
AAH5491906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54919MD Supplier Thermo Scientific Chemicals Supplier No. H54919MD
Only null left

CAS: 776296-12-9 | C10H8BrNO | 238.084 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Notes
Refrigerated (2-4°C)

Chemical Identifiers

CAS 776296-12-9
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.084
MDL Number MFCD16038652
InChI Key YNVIAXULFXXTLV-UHFFFAOYSA-N
Synonym 5-bromo-3-methoxy-quinoline, 5-bromo-3-methyoxy-quinoline, quinoline, 5-bromo-3-methoxy
PubChem CID 58227851
IUPAC Name 5-bromo-3-methoxyquinoline
SMILES COC1=CN=C2C=CC=C(C2=C1)Br

Specifications

Quantity 250 mg
Formula Weight 238.08
Percent Purity 96%
Chemical Name or Material 5-Bromo-3-methoxyquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only