missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-3-indolyl beta-D-Galactopyranoside 98.0+%, TCI America™
$29.83 - $83.89
Chemical Identifiers
| CAS | 97753-82-7 |
|---|---|
| Molecular Formula | C14H16BrNO6 |
| Molecular Weight (g/mol) | 374.19 |
| MDL Number | MFCD00063691 |
| InChI Key | LINMATFDVHBYOS-MBJXGIAVSA-N |
| Synonym | 5-bromo-3-indolyl-beta-d-galactopyranoside, 5-bromo-3-indolyl-b-d-galactopyranoside, bluo-gal, blue-gal, 2s,3r,4s,5r,6r-2-5-bromo-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 5-brig, blue gal, 5-bromo-3-indolyl-beta-galactoside, 5-bromo-3-indolyl beta-d-galactopyranoside, 5-bromo-3-indoxyl-beta-d-galactopyranoside |
| PubChem CID | 126898 |
| IUPAC Name | (2S,3R,4S,5R,6R)-2-[(5-bromo-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
| SMILES | OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C=C23)[C@H](O)[C@@H](O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B347020MG
|
TCI America
B347020MG |
20 mg |
Each for $29.83
|
|
|||||
|
B3470100MG
|
TCI America
B3470100MG |
100 mg |
Each for $83.89
|
|
|||||
Chemical Identifiers
| 97753-82-7 | |
| 374.19 | |
| LINMATFDVHBYOS-MBJXGIAVSA-N | |
| 126898 | |
| OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C=C23)[C@H](O)[C@@H](O)[C@H]1O |
| C14H16BrNO6 | |
| MFCD00063691 | |
| 5-bromo-3-indolyl-beta-d-galactopyranoside, 5-bromo-3-indolyl-b-d-galactopyranoside, bluo-gal, blue-gal, 2s,3r,4s,5r,6r-2-5-bromo-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 5-brig, blue gal, 5-bromo-3-indolyl-beta-galactoside, 5-bromo-3-indolyl beta-d-galactopyranoside, 5-bromo-3-indoxyl-beta-d-galactopyranoside | |
| (2S,3R,4S,5R,6R)-2-[(5-bromo-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
Specifications
| 97753-82-7 | |
| C14H16BrNO6 | |
| 20 mg | |
| LINMATFDVHBYOS-MBJXGIAVSA-N | |
| (2S,3R,4S,5R,6R)-2-[(5-bromo-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 126898 | |
| ≥98.0% (HPLC) | |
| 5-Bromo-3-indolyl beta-D-Galactopyranoside [for Biochemical Research] |
| White | |
| MFCD00063691 | |
| 5-bromo-3-indolyl-beta-d-galactopyranoside, 5-bromo-3-indolyl-b-d-galactopyranoside, bluo-gal, blue-gal, 2s,3r,4s,5r,6r-2-5-bromo-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 5-brig, blue gal, 5-bromo-3-indolyl-beta-galactoside, 5-bromo-3-indolyl beta-d-galactopyranoside, 5-bromo-3-indoxyl-beta-d-galactopyranoside | |
| OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C=C23)[C@H](O)[C@@H](O)[C@H]1O | |
| 374.19 | |
| 374.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer