missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-fluoropyridine-2-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 669066-91-5
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 220.00
MDL Number MFCD07437865
InChI Key JXHUXOAJAYWMHK-UHFFFAOYSA-N
Synonym 5-bromo-3-fluoropicolinic acid, 5-bromo-3-fluoro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo-3-fluoro, 5-bromo-3-fluoropicolinicacid, 5-bromo-2-carboxy-3-fluoropyridine, 5-bromo-3-fluoropyridin-2-carboxylicacid, 2-pyridinecarboxylicacid,5-bromo-3-fluoro, 2-pyridinecarboxylicacid, 5-bromo-3-fluoro, 5-bromo-3-fluoro-pyridine-2-carboxylic acid, 5-bromanyl-3-fluoranyl-pyridine-2-carboxylic acid
PubChem CID 21075872
IUPAC Name 5-bromo-3-fluoropyridine-2-carboxylic acid
SMILES OC(=O)C1=NC=C(Br)C=C1F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC446530050
SDS View Documents
Thermo Scientific Chemicals
446530050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

669066-91-5
220.00
JXHUXOAJAYWMHK-UHFFFAOYSA-N
21075872
OC(=O)C1=NC=C(Br)C=C1F
C6H3BrFNO2
MFCD07437865
5-bromo-3-fluoropicolinic acid, 5-bromo-3-fluoro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo-3-fluoro, 5-bromo-3-fluoropicolinicacid, 5-bromo-2-carboxy-3-fluoropyridine, 5-bromo-3-fluoropyridin-2-carboxylicacid, 2-pyridinecarboxylicacid,5-bromo-3-fluoro, 2-pyridinecarboxylicacid, 5-bromo-3-fluoro, 5-bromo-3-fluoro-pyridine-2-carboxylic acid, 5-bromanyl-3-fluoranyl-pyridine-2-carboxylic acid
5-bromo-3-fluoropyridine-2-carboxylic acid

Specifications

669066-91-5
C6H3BrFNO2
5g
JXHUXOAJAYWMHK-UHFFFAOYSA-N
5-bromo-3-fluoropyridine-2-carboxylic acid
21075872
97%
96% min. (HPLC)
MFCD07437865
5-bromo-3-fluoropicolinic acid, 5-bromo-3-fluoro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo-3-fluoro, 5-bromo-3-fluoropicolinicacid, 5-bromo-2-carboxy-3-fluoropyridine, 5-bromo-3-fluoropyridin-2-carboxylicacid, 2-pyridinecarboxylicacid,5-bromo-3-fluoro, 2-pyridinecarboxylicacid, 5-bromo-3-fluoro, 5-bromo-3-fluoro-pyridine-2-carboxylic acid, 5-bromanyl-3-fluoranyl-pyridine-2-carboxylic acid
OC(=O)C1=NC=C(Br)C=C1F
220.00
220
5-Bromo-3-fluoropyridine-2-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only