missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-fluoropyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. AC446530050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446530050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC446530050 Supplier Thermo Scientific Chemicals Supplier No. 446530050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 669066-91-5
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 220.00
MDL Number MFCD07437865
InChI Key JXHUXOAJAYWMHK-UHFFFAOYSA-N
PubChem CID 21075872
IUPAC Name 5-bromo-3-fluoropyridine-2-carboxylic acid
SMILES OC(=O)C1=NC=C(Br)C=C1F

Specifications

Melting Point 165°C to 170°C
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Synonym 5-bromo-3-fluoropicolinic acid, 5-bromo-3-fluoro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo-3-fluoro, 5-bromo-3-fluoropicolinicacid, 5-bromo-2-carboxy-3-fluoropyridine, 5-bromo-3-fluoropyridin-2-carboxylicacid, 2-pyridinecarboxylicacid,5-bromo-3-fluoro, 2-pyridinecarboxylicacid, 5-bromo-3-fluoro, 5-bromo-3-fluoro-pyridine-2-carboxylic acid, 5-bromanyl-3-fluoranyl-pyridine-2-carboxylic acid
Formula Weight 220
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-fluoropyridine-2-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only