missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-chloro-2-methoxypyridine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 848366-28-9
Molecular Formula C6H5BrClNO
Molecular Weight (g/mol) 222.47
InChI Key XCHXNAQRMMXBGN-UHFFFAOYSA-N
Synonym 2-methoxy-3-chloro-5-bromopyridine, 5-bromo-3-chloro-2-methoxy-pyridine, pyridine, 5-bromo-3-chloro-2-methoxy, 2-methoxyl-3-chloro-5bromopyridine, 3-chloro-2-methoxy-5-bromopyridine, pubchem1164, acmc-209pww, abbypharma ap-11-5379, 2-methoxy-3-chloro-5-bromo-yridine, pyridine,5-bromo-3-chloro-2-methoxy
PubChem CID 40427004
IUPAC Name 5-bromo-3-chloro-2-methoxypyridine
SMILES COC1=NC=C(C=C1Cl)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC449890050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

848366-28-9
222.47
2-methoxy-3-chloro-5-bromopyridine, 5-bromo-3-chloro-2-methoxy-pyridine, pyridine, 5-bromo-3-chloro-2-methoxy, 2-methoxyl-3-chloro-5bromopyridine, 3-chloro-2-methoxy-5-bromopyridine, pubchem1164, acmc-209pww, abbypharma ap-11-5379, 2-methoxy-3-chloro-5-bromo-yridine, pyridine,5-bromo-3-chloro-2-methoxy
5-bromo-3-chloro-2-methoxypyridine
C6H5BrClNO
XCHXNAQRMMXBGN-UHFFFAOYSA-N
40427004
COC1=NC=C(C=C1Cl)Br

Specifications

848366-28-9
C6H5BrClNO
XCHXNAQRMMXBGN-UHFFFAOYSA-N
5-bromo-3-chloro-2-methoxypyridine
40427004
97%
96% min. (GC)
2-methoxy-3-chloro-5-bromopyridine, 5-bromo-3-chloro-2-methoxy-pyridine, pyridine, 5-bromo-3-chloro-2-methoxy, 2-methoxyl-3-chloro-5bromopyridine, 3-chloro-2-methoxy-5-bromopyridine, pubchem1164, acmc-209pww, abbypharma ap-11-5379, 2-methoxy-3-chloro-5-bromo-yridine, pyridine,5-bromo-3-chloro-2-methoxy
COC1=NC=C(C=C1Cl)Br
222.47
222.47
5-Bromo-3-chloro-2-methoxypyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only