missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(trifluoromethyl)pyridine, 97%

Catalog No. AAB2573106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2573106 5 g
AAB2573103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2573106 Supplier Thermo Scientific Chemicals Supplier No. B2573106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 436799-32-5 | C6H3BrF3N | 225.996 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 436799-32-5
Molecular Formula C6H3BrF3N
MDL Number MFCD06657686
InChI Key RPFAUCIXZGMCFN-UHFFFAOYSA-N
Synonym 5-bromo-2-trifluoromethyl pyridine, 2-trifluoromethyl-5-bromopyridine, pyridine, 5-bromo-2-trifluoromethyl, trifluoromethyl-5-bromopyridine, trifluoromethyl-5-bromo-2-pyridine, 5-bromo-2-trifluoromethyl-pyridine, 5-bromo-2 trifluoromethyl pyridine, 5-bromo-2-trifluormethyl pyridine, pubchem2164
PubChem CID 2761197
IUPAC Name 5-bromo-2-(trifluoromethyl)pyridine

Specifications

Melting Point 39°C to 40°C
Boiling Point 76°C to 78°C (18 mmHg)
Quantity 5 g
SMILES C1=CC(=NC=C1Br)C(F)(F)F
Molecular Weight (g/mol) 225.996
Formula Weight 226
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only