missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(trifluoromethoxy)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. p-7030867
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3175109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3175109 Supplier Thermo Scientific Chemicals Supplier No. H3175109

5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Chemical Identifiers

CAS 923281-52-1
Molecular Formula C8H4BrF3O2
Molecular Weight (g/mol) 269.02
MDL Number MFCD03701029
InChI Key IBEWLWBZTHFCPI-UHFFFAOYSA-N
Synonym 5-bromo-2-trifluoromethoxy benzaldehyde, benzaldehyde, 5-bromo-2-trifluoromethoxy, acmc-209rfd, 2-trifluoromethoxy-5-bromobenzaldehyde, 4-bromo-2-formyl-alpha,alpha,alpha-trifluoroanisole
PubChem CID 26599201
IUPAC Name 5-bromo-2-(trifluoromethoxy)benzaldehyde
SMILES FC(F)(F)OC1=C(C=O)C=C(Br)C=C1

Specifications

Quantity 10 g
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 269.01
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-(trifluoromethoxy)benzaldehyde
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only