missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-thiophenemethanol, 95%

Catalog No. AAH6149306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6149306 5 g
AAH6149314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6149306 Supplier Thermo Scientific Chemicals Supplier No. H6149306

CAS: 79387-71-6 | C5H5BrOS | 193.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79387-71-6
Molecular Formula C5H5BrOS
Molecular Weight (g/mol) 193.06
MDL Number MFCD06202626
InChI Key DBRSLMCLMMFHEF-UHFFFAOYSA-N
PubChem CID 11435467
IUPAC Name (5-bromothiophen-2-yl)methanol
SMILES OCC1=CC=C(Br)S1

Specifications

Color Yellow
Quantity 5 g
Synonym 5-bromothien-2-yl methanol, 5-bromothiophen-2-yl methanol, 5-bromo-2-thiophenemethanol, 2-thiophenemethanol, 5-bromo, 5-bromothiophene-2-methanol, 5-bromo-2-thienyl methanol, ksc646o4p, 2-thiophenemethanol,5-bromo, 5-bromo-2-thiophenyl methanol, 5-bromo-2-hydroxymethylthiophene
Formula Weight 193.06
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-thiophenemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only