missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(tetrahydropyran-4-yloxy)pyridine, 97%, Thermo Scientific™

Catalog No. CC58310DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC58310DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC58310DA Supplier Thermo Scientific Chemicals Supplier No. CC58310DA

Chemical Identifiers

CAS 494772-07-5
Molecular Formula C10H12BrNO2
Molecular Weight (g/mol) 258.115
MDL Number MFCD09064985
InChI Key ZCTXTBRJCPHMEP-UHFFFAOYSA-N
Synonym 5-bromo-2-tetrahydro-2h-pyran-4-yloxy pyridine, 5-bromo-2-tetrahydro-2h-pyran-4-yl oxy pyridine, 5-bromo-2-tetrahydropyran-4-yloxy pyridine, 5-bromo-2-oxan-4-yloxy pyridine, pyridine,5-bromo-2-tetrahydro-2h-pyran-4-yl oxy, pubchem22061, 5-bromo-2-oxan-4-yl oxy pyridine, 2-tetrahydropyran-4-yloxy-5-bromopridine, 2-tetrahydropyran-4-yloxy-5-bromopyridine, 4-5-bromopyridin-2-yl oxy oxane
PubChem CID 21914391
IUPAC Name 5-bromo-2-(oxan-4-yloxy)pyridine
SMILES C1COCCC1OC2=NC=C(C=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 258.11
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-(tetrahydropyran-4-yloxy)pyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.