missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-pyridinemethanol 98.0+%, TCI America™
$86.64 - $263.75
Chemical Identifiers
| CAS | 88139-91-7 |
|---|---|
| Molecular Formula | C6H6BrNO |
| Molecular Weight (g/mol) | 188.02 |
| MDL Number | MFCD04035597 |
| InChI Key | RUCZFWMEACWFER-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-hydroxymethylpyridine, 2-hydroxymethyl-5-bromopyridine, 5-bromopyridin-2-yl methanol, 5-bromo-2-hydroxymethyl pyridine, 5-bromopyrid-2-yl methanol, 5-bromo-2-pyridinemethanol, 5-bromo-pyridin-2-yl-methanol, 2-pyridinemethanol, 5-bromo, 5-bromo-2-pyridyl methan-1-ol, 5-bromopyridine-2-methanol |
| PubChem CID | 5200169 |
| IUPAC Name | (5-bromopyridin-2-yl)methanol |
| SMILES | OCC1=CC=C(Br)C=N1 |
Chemical Identifiers
| 88139-91-7 | |
| 188.02 | |
| RUCZFWMEACWFER-UHFFFAOYSA-N | |
| 5200169 | |
| OCC1=CC=C(Br)C=N1 |
| C6H6BrNO | |
| MFCD04035597 | |
| 5-bromo-2-hydroxymethylpyridine, 2-hydroxymethyl-5-bromopyridine, 5-bromopyridin-2-yl methanol, 5-bromo-2-hydroxymethyl pyridine, 5-bromopyrid-2-yl methanol, 5-bromo-2-pyridinemethanol, 5-bromo-pyridin-2-yl-methanol, 2-pyridinemethanol, 5-bromo, 5-bromo-2-pyridyl methan-1-ol, 5-bromopyridine-2-methanol | |
| (5-bromopyridin-2-yl)methanol |
Specifications
| 88139-91-7 | |
| 120°C | |
| MFCD04035597 | |
| 5-bromo-2-hydroxymethylpyridine, 2-hydroxymethyl-5-bromopyridine, 5-bromopyridin-2-yl methanol, 5-bromo-2-hydroxymethyl pyridine, 5-bromopyrid-2-yl methanol, 5-bromo-2-pyridinemethanol, 5-bromo-pyridin-2-yl-methanol, 2-pyridinemethanol, 5-bromo, 5-bromo-2-pyridyl methan-1-ol, 5-bromopyridine-2-methanol | |
| OCC1=CC=C(Br)C=N1 | |
| 188.02 | |
| 188.02 | |
| Crystalline Powder |
| 61°C | |
| C6H6BrNO | |
| 1 g | |
| RUCZFWMEACWFER-UHFFFAOYSA-N | |
| (5-bromopyridin-2-yl)methanol | |
| 5200169 | |
| ≥98.0% (GC,T) | |
| 5-Bromo-2-pyridinemethanol |
Safety and Handling
TSCA : No