missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-pyridinemethanol 98.0+%, TCI America™

Catalog No. B34921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34921G 1 g
B34925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34921G Supplier TCI America Supplier No. B34921G
Only null left

Chemical Identifiers

CAS 88139-91-7
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD04035597
InChI Key RUCZFWMEACWFER-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxymethylpyridine, 2-hydroxymethyl-5-bromopyridine, 5-bromopyridin-2-yl methanol, 5-bromo-2-hydroxymethyl pyridine, 5-bromopyrid-2-yl methanol, 5-bromo-2-pyridinemethanol, 5-bromo-pyridin-2-yl-methanol, 2-pyridinemethanol, 5-bromo, 5-bromo-2-pyridyl methan-1-ol, 5-bromopyridine-2-methanol
PubChem CID 5200169
IUPAC Name (5-bromopyridin-2-yl)methanol
SMILES OCC1=CC=C(Br)C=N1

Specifications

Melting Point 61°C
Boiling Point 120°C
Quantity 1 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-pyridinemethanol