missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-pyridinecarboxylic Acid 97.0+%, TCI America™

Catalog No. p-7138111
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31881G 1 g
B31885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31881G Supplier TCI America Supplier No. B31881G
Only null left

Chemical Identifiers

CAS 30766-11-1
Molecular Formula C6H3BrNO2
Molecular Weight (g/mol) 201.00
MDL Number MFCD00234149
InChI Key MNNQIBXLAHVDDL-UHFFFAOYSA-M
Synonym 5-bromopicolinic acid, 5-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo, 5-bromo-2-carboxypyridine, 5-bromo-pyridine-2-carboxylic acid, 5-bromo-2-picolinic acid, 5-bromo-2-carboxy pyridine, 5-bromo-2-pyridine carboxylic acid, 5-bromopyridine-2-carboxylicacid
PubChem CID 608544
IUPAC Name 5-bromopyridine-2-carboxylate
SMILES [O-]C(=O)C1=CC=C(Br)C=N1

Specifications

Melting Point 176°C
Quantity 1 g
Formula Weight 202.01
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-pyridinecarboxylic Acid