missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. B34891G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34891G Supplier TCI America Supplier No. B34891G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31181-90-5
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
MDL Number MFCD04112535
InChI Key ZQVLPMNLLKGGIU-UHFFFAOYSA-N
Synonym 5-bromopicolinaldehyde, 5-bromo-2-formylpyridine, 5-bromo-2-pyridinecarboxaldehyde, 5-bromopyridine-2-carboxaldehyde, 5-bromo-pyridine-2-carbaldehyde, 5-bromo-2-pyridinecarbaldehyde, 2-pyridinecarboxaldehyde, 5-bromo, 5-bromo-2-pyridine-carbaldehyde, pubchem5059, 5-bromo picolinaldehyde
PubChem CID 9877562
IUPAC Name 5-bromopyridine-2-carbaldehyde
SMILES C1=CC(=NC=C1Br)C=O

Specifications

Melting Point 96°C
Boiling Point 70°C
Quantity 1 g
Formula Weight 186.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-pyridinecarboxaldehyde
TSCA No
Recommended Storage Refrigerator