missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-nitropyridine 98.0+%, TCI America™

Catalog No. B27661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27661G 1 g
B27665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27661G Supplier TCI America Supplier No. B27661G

Chemical Identifiers

CAS 39856-50-3
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 203.00
MDL Number MFCD00160411
InChI Key ATXXLNCPVSUCNK-UHFFFAOYSA-N
Synonym 2-nitro-5-bromopyridine, pyridine, 5-bromo-2-nitro, 2-nitro-5-bromo pyridine, 5-bromo-2-nitro-pyridine, 5-brom-2-nitropyridin, pubchem6691, 5-bromo2-nitropyridine, 5-bromo-2-nitropyridin, 3-bromo-6-nitro pyridine, 5-bromo-2-nitro pyridine
PubChem CID 817620
IUPAC Name 5-bromo-2-nitropyridine
SMILES [O-][N+](=O)C1=CC=C(Br)C=N1

Specifications

Melting Point 150°C
Quantity 1 g
Formula Weight 203.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-nitropyridine