missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-nitropyridine, 98+%, Thermo Scientific Chemicals

Catalog No. AAL1974806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1974806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1974806 Supplier Thermo Scientific Chemicals Supplier No. L1974806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39856-50-3 | Molecular Formula: C5H3BrN2O2 | Molecular Weight: 203.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39856-50-3
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 203.00
MDL Number MFCD00160411
InChI Key ATXXLNCPVSUCNK-UHFFFAOYSA-N
Synonym 2-nitro-5-bromopyridine, pyridine, 5-bromo-2-nitro, 2-nitro-5-bromo pyridine, 5-bromo-2-nitro-pyridine, 5-brom-2-nitropyridin, pubchem6691, 5-bromo2-nitropyridine, 5-bromo-2-nitropyridin, 3-bromo-6-nitro pyridine, 5-bromo-2-nitro pyridine
PubChem CID 817620
IUPAC Name 5-bromo-2-nitropyridine
SMILES [O-][N+](=O)C1=CC=C(Br)C=N1

Specifications

Melting Point 147°C to 151°C
Quantity 5 g
Beilstein 120878
Formula Weight 203
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-nitropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only