missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(methylthio)pyrimidine-4-carboxylic Acid 97.0+%, TCI America™

Catalog No. B38291G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38291G 1 g
B38295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38291G Supplier TCI America Supplier No. B38291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50593-92-5
Molecular Formula C6H5BrN2O2S
Molecular Weight (g/mol) 249.08
MDL Number MFCD00238866
InChI Key YJEWVVYJOJJLBP-UHFFFAOYSA-N
Synonym 5-bromo-2-methylthio pyrimidine-4-carboxylic acid, 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylic acid, 5-bromo-2-methylsulfanyl pyrimidine-4-carboxylic acid, 4-pyrimidinecarboxylic acid, 5-bromo-2-methylthio, 5-bromo-2-methylsulfanyl-pyrimidine-4-carboxylic acid, 5-bromo-2-methylthio pyrimidine-4-carboxylicacid, 5-bromo-2-methylthio-4-pyrimidinecarboxylic acid, 5-bromo-2-methylsulphanyl pyrimidine-4-carboxylic acid, pubchem13048
PubChem CID 291786
IUPAC Name 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylic acid
SMILES CSC1=NC=C(Br)C(=N1)C(O)=O

Specifications

Melting Point 177°C
Quantity 1 g
Formula Weight 249.08
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-(methylthio)pyrimidine-4-carboxylic Acid