missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylpyrimidine 98.0+%, TCI America™

Catalog No. B52661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52661G 1 g
B52665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52661G Supplier TCI America Supplier No. B52661G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7752-78-5
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD07375143
InChI Key NEDJTEXNSTUKHW-UHFFFAOYSA-N
Synonym 5-bromo-2-methyl-pyrimidine, 2-methyl-5-bromopyrimidine, pyrimidine, 5-bromo-2-methyl, 5-bromo-2-methyl-1,3-diazine, pubchem15231, acmc-1bm3h, 5-bromo-2-methyl pyrimidine
PubChem CID 14387744
IUPAC Name 5-bromo-2-methylpyrimidine
SMILES CC1=NC=C(C=N1)Br

Specifications

Melting Point 83°C
Quantity 1 g
Formula Weight 173.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methylpyrimidine