missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylpyrimidine, 98%

Catalog No. p-7057111
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66467MD 250 mg
AAH6646703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66467MD Supplier Thermo Scientific Chemicals Supplier No. H66467MD

CAS: 7752-78-5 | C5H5BrN2 | 173.013 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7752-78-5
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD07375143
InChI Key NEDJTEXNSTUKHW-UHFFFAOYSA-N
Synonym 5-bromo-2-methyl-pyrimidine, 2-methyl-5-bromopyrimidine, pyrimidine, 5-bromo-2-methyl, 5-bromo-2-methyl-1,3-diazine, pubchem15231, acmc-1bm3h, 5-bromo-2-methyl pyrimidine
PubChem CID 14387744
IUPAC Name 5-bromo-2-methylpyrimidine
SMILES CC1=NC=C(C=N1)Br

Specifications

Quantity 250 mg
Beilstein 108736á
Formula Weight 173.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-methylpyrimidine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only