missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxypyrimidine 98.0+%, TCI America™

Catalog No. B35211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35211G 1 g
B35215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35211G Supplier TCI America Supplier No. B35211G

Chemical Identifiers

CAS 14001-66-2
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.012
MDL Number MFCD04973351
InChI Key DWVCZDMMGYIULX-UHFFFAOYSA-N
Synonym 2-methoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2-methoxy, 5-bromo-2-methoxy-pyrimidine, 5-bromo-2-methoxy-1,3-diazine, 5-bromopyrimidin-2-yl methyl ether, 5-bromo-2-methyloxy pyrimidine, pubchem5264, acmc-209cjt, 5-bromo-2methoxypyrimidine, ksc494g8n
PubChem CID 139657
IUPAC Name 5-bromo-2-methoxypyrimidine
SMILES COC1=NC=C(C=N1)Br

Specifications

Melting Point 56°C
Color White-Yellow
Quantity 1 g
Formula Weight 189.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxypyrimidine