missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxypyrimidine, 97%

Catalog No. AC441850010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441850010 1 g
AC441850050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC441850010 Supplier Thermo Scientific Chemicals Supplier No. 441850010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14001-66-2 | C5H5BrN2O | 189.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14001-66-2
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.01
InChI Key DWVCZDMMGYIULX-UHFFFAOYSA-N
Synonym 2-methoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2-methoxy, 5-bromo-2-methoxy-pyrimidine, 5-bromo-2-methoxy-1,3-diazine, 5-bromopyrimidin-2-yl methyl ether, 5-bromo-2-methyloxy pyrimidine, pubchem5264, acmc-209cjt, 5-bromo-2methoxypyrimidine, ksc494g8n
PubChem CID 139657
IUPAC Name 5-bromo-2-methoxypyrimidine
SMILES COC1=NC=C(C=N1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 57°C to 59°C
Color Pink-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 189.01
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-methoxypyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only