missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxypyridine 98.0+%, TCI America™

Catalog No. p-7119654
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23985G 5 g
B239825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23985G Supplier TCI America Supplier No. B23985G
Only null left

Chemical Identifiers

CAS 13472-85-0
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
InChI Key XADICJHFELMBGX-UHFFFAOYSA-N
Synonym 2-methoxy-5-bromopyridine, pyridine, 5-bromo-2-methoxy, 3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-pyridine, 2-methyloxy-5-bromopyridine, 5-bromo-2-methoxy pyridine, 2-methoxy-5-bromo-pyridine, 2-methyloxy-5-bromo pyridine, zlchem 355, pubchem2408
PubChem CID 2734895
IUPAC Name 5-bromo-2-methoxypyridine
SMILES COC1=NC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 195°C
Quantity 5 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-methoxypyridine