missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxybenzenesulfonyl Chloride 97.0+%, TCI America™

Catalog No. B39651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39651G 1 g
B39655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39651G Supplier TCI America Supplier No. B39651G

Chemical Identifiers

CAS 23095-05-8
Molecular Formula C7H6BrClO3S
Molecular Weight (g/mol) 285.536
MDL Number MFCD00051768
InChI Key IXSBNNRUQYYMRM-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxybenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 5-bromo-2-methoxy, 5-bromo-2-methoxybenzenesulphonyl chloride, 5-bromo-2-methoxy-benzenesulfonyl chloride, pubchem16355, acmc-1clw8, ksc495e9j, buttpark 100\07-65, 5-bromo-2-methoxyphenyl chlorosulfone, 5-bromo-2methoxybenzenesulfonyl chloride
PubChem CID 520020
IUPAC Name 5-bromo-2-methoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)Br)S(=O)(=O)Cl

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 285.54
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxybenzenesulfonyl Chloride
TSCA No
Recommended Storage Refrigerator