missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxybenzenesulfonyl chloride, 98%

Catalog No. p-7021806
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1429314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1429314 Supplier Thermo Scientific Chemicals Supplier No. A1429314
Only null left

CAS: 23095-05-8 | C7H6BrClO3S | 285.536 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23095-05-8
Molecular Formula C7H6BrClO3S
Molecular Weight (g/mol) 285.536
MDL Number MFCD00051768
InChI Key IXSBNNRUQYYMRM-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxybenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 5-bromo-2-methoxy, 5-bromo-2-methoxybenzenesulphonyl chloride, 5-bromo-2-methoxy-benzenesulfonyl chloride, pubchem16355, acmc-1clw8, ksc495e9j, buttpark 100\07-65, 5-bromo-2-methoxyphenyl chlorosulfone, 5-bromo-2methoxybenzenesulfonyl chloride
PubChem CID 520020
IUPAC Name 5-bromo-2-methoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)Br)S(=O)(=O)Cl

Specifications

Melting Point 112°C to 115°C
Quantity 25 g
UN Number UN3261
Beilstein 3093014
Sensitivity Moisture sensitive
Formula Weight 285.54
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-methoxybenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only