missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-methoxy-3-nitropyridine 96.0+%, TCI America™
$65.24 - $212.51
Chemical Identifiers
| CAS | 152684-30-5 |
|---|---|
| Molecular Formula | C6H5BrN2O3 |
| Molecular Weight (g/mol) | 233.02 |
| MDL Number | MFCD07374968 |
| InChI Key | YRVHFGOAEVWBNS-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-methoxy-3-nitro-pyridine, 5-bromo-2-methyloxy-3-nitropyridine, pyridine, 5-bromo-2-methoxy-3-nitro, pubchem6589, ksc498c2d, acmc-1c049, 2-methoxy-3-nitro-5-bromopyridine, abbypharma ap-11-5930, 2-methoxy-3-nitro-5-bromo pyridine, 5-bromanyl-2-methoxy-3-nitro-pyridine |
| PubChem CID | 15645664 |
| IUPAC Name | 5-bromo-2-methoxy-3-nitropyridine |
| SMILES | COC1=NC=C(Br)C=C1[N+]([O-])=O |
Chemical Identifiers
| 152684-30-5 | |
| 233.02 | |
| YRVHFGOAEVWBNS-UHFFFAOYSA-N | |
| 15645664 | |
| COC1=NC=C(Br)C=C1[N+]([O-])=O |
| C6H5BrN2O3 | |
| MFCD07374968 | |
| 5-bromo-2-methoxy-3-nitro-pyridine, 5-bromo-2-methyloxy-3-nitropyridine, pyridine, 5-bromo-2-methoxy-3-nitro, pubchem6589, ksc498c2d, acmc-1c049, 2-methoxy-3-nitro-5-bromopyridine, abbypharma ap-11-5930, 2-methoxy-3-nitro-5-bromo pyridine, 5-bromanyl-2-methoxy-3-nitro-pyridine | |
| 5-bromo-2-methoxy-3-nitropyridine |
Specifications
| 152684-30-5 | |
| C6H5BrN2O3 | |
| 1 g | |
| YRVHFGOAEVWBNS-UHFFFAOYSA-N | |
| 5-bromo-2-methoxy-3-nitropyridine | |
| 15645664 | |
| ≥96.0% (GC) | |
| 5-Bromo-2-methoxy-3-nitropyridine |
| 90°C | |
| MFCD07374968 | |
| 5-bromo-2-methoxy-3-nitro-pyridine, 5-bromo-2-methyloxy-3-nitropyridine, pyridine, 5-bromo-2-methoxy-3-nitro, pubchem6589, ksc498c2d, acmc-1c049, 2-methoxy-3-nitro-5-bromopyridine, abbypharma ap-11-5930, 2-methoxy-3-nitro-5-bromo pyridine, 5-bromanyl-2-methoxy-3-nitro-pyridine | |
| COC1=NC=C(Br)C=C1[N+]([O-])=O | |
| 233.02 | |
| 233.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : No