missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxy-3-nitropyridine 96.0+%, TCI America™

Catalog No. B44615G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44615G 5 g
B44611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44615G Supplier TCI America Supplier No. B44615G

Chemical Identifiers

CAS 152684-30-5
Molecular Formula C6H5BrN2O3
Molecular Weight (g/mol) 233.02
MDL Number MFCD07374968
InChI Key YRVHFGOAEVWBNS-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxy-3-nitro-pyridine, 5-bromo-2-methyloxy-3-nitropyridine, pyridine, 5-bromo-2-methoxy-3-nitro, pubchem6589, ksc498c2d, acmc-1c049, 2-methoxy-3-nitro-5-bromopyridine, abbypharma ap-11-5930, 2-methoxy-3-nitro-5-bromo pyridine, 5-bromanyl-2-methoxy-3-nitro-pyridine
PubChem CID 15645664
IUPAC Name 5-bromo-2-methoxy-3-nitropyridine
SMILES COC1=NC=C(Br)C=C1[N+]([O-])=O

Specifications

Melting Point 90°C
Quantity 5 g
Formula Weight 233.02
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxy-3-nitropyridine