missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxy-3-methylpyridine, 97%

Catalog No. p-7033101
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5011303 1 g
AAH5011306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5011303 Supplier Thermo Scientific Chemicals Supplier No. H5011303
Only null left

CAS: 760207-87-2 | C7H8BrNO | 202.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 760207-87-2
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD07375124
InChI Key RHYCFMXPPGZHAW-UHFFFAOYSA-N
Synonym pyridine, 5-bromo-2-methoxy-3-methyl, 5-bromo-3-methyl-2-methoxypyridine, 5-bromo-2-methoxy-3-methyl-pyridine, 5-bromo-2-methoxy-3-picoline, pubchem22058, acmc-209p1f, ksc496k0b, abbypharma ap-10-5114
PubChem CID 22591176
IUPAC Name 5-bromo-2-methoxy-3-methylpyridine
SMILES CC1=CC(=CN=C1OC)Br

Specifications

Refractive Index 1.554
Quantity 1 g
Formula Weight 202.05
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methoxy-3-methylpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only