missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-isopropoxypyridine, 97%

Catalog No. p-7031476
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3256403 1 g
AAH3256406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3256403 Supplier Thermo Scientific Chemicals Supplier No. H3256403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 870521-31-6 | C8H10BrNO | 216.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 870521-31-6
Molecular Formula C8H10BrNO
Molecular Weight (g/mol) 216.08
MDL Number MFCD07368869
InChI Key FSZRSTNRZUIEMA-UHFFFAOYSA-N
Synonym 5-bromo-2-isopropoxypyridine, 5-bromo-2-propan-2-yloxy pyridine, 5-bromo-2-isopropoxy-pyridine, 3-bromo-6-i-propoxy pyridine, pyridine, 5-bromo-2-1-methylethoxy, pubchem22060, acmc-209qd4, 2-isopropoxy-5-bromo-pyridine, 5-bromo-2-iso-propoxypyridine, 5-bromo-2-isopropyloxypyridine
PubChem CID 21935068
IUPAC Name 5-bromo-2-propan-2-yloxypyridine
SMILES CC(C)OC1=NC=C(Br)C=C1

Specifications

Refractive Index 1.5305
Quantity 1 g
Formula Weight 216.08
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-isopropoxypyridine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only