missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-iodopyridine 97.0+%, TCI America™
$78.93 - $309.95
Chemical Identifiers
| CAS | 223463-13-6 |
|---|---|
| Molecular Formula | C5H3BrIN |
| Molecular Weight (g/mol) | 283.894 |
| MDL Number | MFCD00691506 |
| InChI Key | HSNBRDZXJMPDGH-UHFFFAOYSA-N |
| Synonym | 2-iodo-5-bromopyridine, 5-bromo-2-iodo-pyridine, 2-iodo-5-bromo-pyridine, 2-iodo-5-bromo pyridine, pyridine, 5-bromo-2-iodo, 5-bromo-2-iodo pyridine, pubchem2409, 5-cromo-2-iodopyridine, 5-bromo-2-iodopyridine, acmc-209fv4 |
| PubChem CID | 593690 |
| IUPAC Name | 5-bromo-2-iodopyridine |
| SMILES | C1=CC(=NC=C1Br)I |
Chemical Identifiers
| 223463-13-6 | |
| 283.894 | |
| HSNBRDZXJMPDGH-UHFFFAOYSA-N | |
| 593690 | |
| C1=CC(=NC=C1Br)I |
| C5H3BrIN | |
| MFCD00691506 | |
| 2-iodo-5-bromopyridine, 5-bromo-2-iodo-pyridine, 2-iodo-5-bromo-pyridine, 2-iodo-5-bromo pyridine, pyridine, 5-bromo-2-iodo, 5-bromo-2-iodo pyridine, pubchem2409, 5-cromo-2-iodopyridine, 5-bromo-2-iodopyridine, acmc-209fv4 | |
| 5-bromo-2-iodopyridine |
Specifications
| 223463-13-6 | |
| White-Yellow | |
| MFCD00691506 | |
| 2-iodo-5-bromopyridine, 5-bromo-2-iodo-pyridine, 2-iodo-5-bromo-pyridine, 2-iodo-5-bromo pyridine, pyridine, 5-bromo-2-iodo, 5-bromo-2-iodo pyridine, pubchem2409, 5-cromo-2-iodopyridine, 5-bromo-2-iodopyridine, acmc-209fv4 | |
| C1=CC(=NC=C1Br)I | |
| 283.894 | |
| 283.89 | |
| Crystalline Powder |
| 115°C | |
| C5H3BrIN | |
| 5 g | |
| HSNBRDZXJMPDGH-UHFFFAOYSA-N | |
| 5-bromo-2-iodopyridine | |
| 593690 | |
| ≥97.0% (GC) | |
| 5-Bromo-2-iodopyridine |
Safety and Handling
TSCA : No